missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | EphB4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | H1 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:1000-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human EphB4 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?