missing translation for 'onlineSavingsMsg'
Learn More

ent-Voriconazole, TRC

Product Code. 30450198
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450198 50 mg 50mg
30449805 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30450198 Supplier Toronto Research Chemicals Supplier No. TRCV76000550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137234-63-0
Molecular Formula C16 H14 F3 N5 O
Molecular Weight (g/mol) 349.31
Synonym 4-Pyrimidineethanol, ÃŽ±-(2,4-difluorophenyl)-5-fluoro-ÃŽ2-methyl-ÃŽ±-(1H-1,2,4-triazol-1-ylmethyl)-, (ÃŽ±S,ÃŽ2R)-, 4-Pyrimidineethanol, ÃŽ±-(2,4-difluorophenyl)-5-fluoro-ÃŽ2-methyl-ÃŽ±-(1H-1,2,4-triazol-1-ylmethyl)-, [S-(R*,S*)]-, Voriconazole Imp. D (EP), Voriconazole Related Compound B (USP)
IUPAC Name (2S,3R)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES C[C@H](c1ncncc1F)[C@@](O)(Cn2cncn2)c3ccc(F)cc3F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25H,6H2,1H3/t10-,16+/m1/s1
Formula Weight 349.115
Chemical Name or Material ent-Voriconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.