CAS RN 137234-63-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ent-Voriconazole, TRC
CAS: 137234-63-0 Molecular Formula: C16 H14 F3 N5 O Molecular Weight (g/mol): 349.31 Synonym: 4-Pyrimidineethanol, ÃŽ±-(2,4-difluorophenyl)-5-fluoro-ÃŽ2-methyl-ÃŽ±-(1H-1,2,4-triazol-1-ylmethyl)-, (ÃŽ±S,ÃŽ2R)-,4-Pyrimidineethanol, ÃŽ±-(2,4-difluorophenyl)-5-fluoro-ÃŽ2-methyl-ÃŽ±-(1H-1,2,4-triazol-1-ylmethyl)-, [S-(R*,S*)]-,Voriconazole Imp. D (EP),Voriconazole Related Compound B (USP) IUPAC Name: (2S,3R)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol SMILES: C[C@H](c1ncncc1F)[C@@](O)(Cn2cncn2)c3ccc(F)cc3F