missing translation for 'onlineSavingsMsg'
Learn More

Endothelin-1 Antibody (TR.ET.48.5), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18215100 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215100 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215100 Supplier Novus Biologicals Supplier No. NB300526AF405
  • ONLINE EXCLUSIVE PRICE
    674.00 EUR
    637.35 EUR / 0.1mL
    Save 36.65 EUR
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endothelin-1 Monoclonal specifically detects Endothelin-1 in Canine, Human, Mouse, Pig, Rabbit, Rat, Sheep samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endothelin-1
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone TR.ET.48.5
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P05305
Gene Alias endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1
Gene Symbols EDN1
Host Species Mouse
Immunogen Pure endothelin-1 conjugated to KLH.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1906
Test Specificity Not yet tested in other species.
Target Species Canine, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.