Learn More
Emoxipine, TRC
Product Code. 30135915
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135915 5 g 5g
30007903 500 mg 500mg
30011581 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135915 Supplier Toronto Research Chemicals Supplier No. TRCE5240005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2364-75-2
Molecular Formula C8 H11 N O
Molecular Weight (g/mol) 137.18
Synonym 3-Pyridinol, 2-ethyl-6-methyl-, 2-Ethyl-6-methyl-3-pyridinol, 2-Ethyl-3-hydroxy-6-methylpyridine, 2-Ethyl-6-methyl-3-hydroxypyridine, 6-Methyl-2-ethyl-3-hydroxypyridine, Emoxibel, Emoxipin, Emoxipine, Emoxypin, Emoxypine, Epigid, Epygid
IUPAC Name 2-ethyl-6-methylpyridin-3-ol
SMILES CCc1nc(C)ccc1O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H11NO/c1-3-7-8(10)5-4-6(2)9-7/h4-5,10H,3H2,1-2H3
Formula Weight 137.0841
Percent Purity >95%
Chemical Name or Material Emoxipine
Grade HPLC