accessibility menu, dialog, popup

UserName

HClH3CNCH3HO

CAS RN 2364-75-2

IUPAC Name: hydrogen 2-ethyl-6-methylpyridin-3-ol chloride
Synonyms: emoxypine
Molecular Weight (g/mol): 173.64
Molecular Formula: C8H12ClNO
InChi Key: KZUIXWYHQJZUOK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCC1=NC(C)=CC=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results