Learn More
STEMCELL Technologies EasySep™ Rat B Cell Isolation Kit
Product Code. 17107941
Encompass
Change view
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17107941 1 x 109 cells 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17107941 Supplier STEMCELL Technologies Supplier No. 19644
Immunomagnetic isolation of untouched rat B cells
  • The EasySep™ Rat B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
Cell Type B Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells