missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit
Description
- The EasySep™ Mouse MDSC (CD11b+Gr1+) Isolation Kit is designed to isolate CD11b+Gr1+ myeloid-derived suppressor cells (MDSCs) from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies directed against non-MDSCs and streptavidin-coated magnetic particles (RapidSpheres™)
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
- Isolated cells are immediately available for downstream applications such as flow cytometry, cell sorting, and cell culture
Specifications
Specifications
| Cell Type | Granulocytes and Subsets, Monocytes, Myeloid Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?