missing translation for 'onlineSavingsMsg'
Learn More

(E)-alpha-(4-Chlorophenyl)cinnamonitrile, 99%, Thermo Scientific™

Product Code. 11431667
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431667 25 g 25g
11421667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431667 Supplier Thermo Scientific Alfa Aesar Supplier No. B25644.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16610-81-4
Molecular Formula C15H10ClN
Molecular Weight (g/mol) 239.702
MDL Number MFCD00082873
InChI Key JJGLLZWUWIKTAG-UVTDQMKNSA-N
Synonym a-p-chlorophenyl cinnamonitrile, e-alpha-4-chlorophenyl cinnamonitrile, e-2-4-chlorophenyl-3-phenylacrylonitrile, 2-4-chlorophenyl-3-phenylacrylonitrile, 2e-2-4-chlorophenyl-3-phenylprop-2-enenitrile, alpha-p-chlorophenyl cinnamonitrile, e-3-phenyl-2-4-chlorophenyl acrylonitrile, e-2-4-chlorophenyl-3-phenylprop-2-enenitrile
PubChem CID 736231
IUPAC Name (E)-2-(4-chlorophenyl)-3-phenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C=C(C#N)C2=CC=C(C=C2)Cl

Specifications

Melting Point 113°C to 118°C
Odor Odorless
Quantity 25 g
Beilstein 2331785
Formula Weight 239.7
Percent Purity 99%
Chemical Name or Material (E)-alpha-(4-Chlorophenyl)cinnamonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.