accessibility menu, dialog, popup

UserName

ClN(Z)

CAS RN 16610-81-4

IUPAC Name: (2Z)-2-(4-chlorophenyl)-3-phenylprop-2-enenitrile
Synonyms: (2Z)-2-(4-chlorophenyl)-3-phenylprop-2-enenitrile
Molecular Weight (g/mol): 239.7
Molecular Formula: C15H10ClN
InChi Key: JJGLLZWUWIKTAG-GXDHUFHOSA-N
SMILES: ClC1=CC=C(C=C1)C(=C\C1=CC=CC=C1)\C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results