missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-(2-((6-(2-Cyanophenoxy)pyrimidin-4-yl)oxy)phenyl)-3-methoxyacrylic Acid, TRC

Product Code. 30133148
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133148 5 mg 5mg
30134591 1 mg 1mg
30006383 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133148 Supplier Toronto Research Chemicals Supplier No. TRCR0702405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185255-09-7
Molecular Formula C21 H15 N3 O5
Molecular Weight (g/mol) 389.36
Synonym Azoxystrobin (free acid), Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, (αE)-, (E)-2-(2-(6-(2-cyano-phenoxy)pyrimidin-4-yloxy)-phenyl)-3-methoxy-acrylic acid
IUPAC Name (E)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoic acid
SMILES CO\C=C(\C(=O)O)/c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H15N3O5/c1-27-12-16(21(25)26)15-7-3-5-9-18(15)29-20-10-19(23-13-24-20)28-17-8-4-2-6-14(17)11-22/h2-10,12-13H,1H3,(H,25,26)/b16-12+
Formula Weight 389.1012
Percent Purity >95%
Chemical Name or Material Azoxystrobin (free acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.