CAS RN 1185255-09-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-2-(2-((6-(2-Cyanophenoxy)pyrimidin-4-yl)oxy)phenyl)-3-methoxyacrylic Acid, TRC
CAS: 1185255-09-7 Molecular Formula: C21 H15 N3 O5 Molecular Weight (g/mol): 389.36 Synonym: Azoxystrobin (free acid),Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, (αE)-,(E)-2-(2-(6-(2-cyano-phenoxy)pyrimidin-4-yloxy)-phenyl)-3-methoxy-acrylic acid IUPAC Name: (E)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoic acid SMILES: CO\C=C(\C(=O)O)/c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2