missing translation for 'onlineSavingsMsg'
Learn More

Drometrizole, TRC

Product Code. 30138673
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138673 25 g 25g
30138332 10 g 10g
17961655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138673 Supplier Toronto Research Chemicals Supplier No. TRCD67940025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2440-22-4
Molecular Formula C13 H11 N3 O
Molecular Weight (g/mol) 225.25
Synonym p-Cresol, 2-(2H-benzotriazol-2-yl)- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4-methylphenol, 2-(2-Hydroxy-5-methyl)benzotriazole, 2-(2-Hydroxy-5-methylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-5-methylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-p-cresol, 2-(2'-Hydroxy-5'-methylphenyl)benzotriazole, 2-(5-Methyl-2-hydroxyphenyl)benzotriazole, 2-(5'-Methyl-2'-hydroxyphenyl)benzotriazole, 2-Benzotriazol-2-yl-4-methylphenol, 4-Methyl-2-(2H-benzotriazol-2-yl)phenol, ADK Stab LA 32, ADK-ARKLS DN 13, BT 1, BT 1 (light stabilizer), Benazol II, Benazol P, Chisorb P, Dainsorb T 1, Drometrizole, Eversorb 71, JCK-W-UA, JF 77, JF 77P, JF 77T, Kemisorb 71, LA 32, Lowilite 55, Mark LA 32, NSC 91885, Seesorb 701, Seikalizer AZ, Solartex UVP, Sumisorb 200, Tinuvin P, Tinuvin PED, Tinuvin UV-P, UV Absorber 1, UV-P, UV-P (UV stabilizer), UVA-P, Uvasorb SV, Uvinul 3033P, Viosorb 520
IUPAC Name 2-(benzotriazol-2-yl)-4-methylphenol
SMILES Cc1ccc(O)c(c1)n2nc3ccccc3n2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H11N3O/c1-9-6-7-13(17)12(8-9)16-14-10-4-2-3-5-11(10)15-16/h2-8,17H,1H3
Formula Weight 225.0902
Chemical Name or Material Drometrizole 2-(2H-Benzotriazol-2-yl)-p-cresol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.