accessibility menu, dialog, popup

UserName

CAS RN 2440-22-4

H3COHNNN

CAS RN 2440-22-4

IUPAC Name: 2-(2H-1,2,3-benzotriazol-2-yl)-4-methylphenol
Synonyms: drometrizole tin P 甲酚曲唑 tinuvin II tinuvin P drometrizol porex P UV absorber-1 benazol II drometrizolum
Molecular Weight (g/mol): 225.25
Molecular Formula: C13H11N3O
InChi Key: MCPKSFINULVDNX-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=C1)N1N=C2C=CC=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results