Learn More
Dodecamethylpentasiloxane, TRC
Product Code. 30137554
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137554 10 mL 10mL
17917464 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137554 Supplier Toronto Research Chemicals Supplier No. TRCD49452510ML
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-63-9
Molecular Formula C12 H36 O4 Si5
Molecular Weight (g/mol) 384.84
Synonym Pentasiloxane, 1,1,1,3,3,5,5,7,7,9,9,9-dodecamethyl- (ACI), 1,1,1,3,3,5,5,7,7,9,9,9-Dodecamethylpentasiloxane (ACI), Pentasiloxane, dodecamethyl- (6CI, 7CI, 8CI, 9CI), DC 200 Fluid 2, Dodecamethylpentasiloxane, KF 96L2CS, MD 3M, PSF 2CST, PSF 2Cst Pure Silicone Fluid, Pentasiloxane, 1,1,1,3,3,5,5,7,7,9,9,9-dodecamethyl- (ACI), 1,1,1,3,3,5,5,7,7,9,9,9-Dodecamethylpentasiloxane (ACI), 1,1,1,3,3,5,5,7,7,9,9,9-Dodecamethylpentasiloxane, Pentasiloxane, dodecamethyl- (6CI, 7CI, 8CI, 9CI), DC 200 Fluid 2, KF 96L2CS, MD 3M, PSF 2CST, PSF 2Cst Pure Silicone Fluid
IUPAC Name bis[[dimethyl(trimethylsilyloxy)silyl]oxy]-dimethylsilane
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
Specifications
Quantity 10 mL
InChI Formula InChI=1S/C12H36O4Si5/c1-17(2,3)13-19(7,8)15-21(11,12)16-20(9,10)14-18(4,5)6/h1-12H3
Formula Weight 384.146
Chemical Name or Material Dodecamethylpentasiloxane