accessibility menu, dialog, popup

UserName

CAS RN 141-63-9

CH3SiCH3H3COSiCH3H3COSiCH3H3COSiCH3H3COSiCH3CH3H3C

CAS RN 141-63-9

IUPAC Name: 2,2,4,4,6,6,8,8,10,10-decamethyl-3,5,7,9-tetraoxa-2,4,6,8,10-pentasilaundecane
Synonyms: dodecamethylpentasiloxane
Molecular Weight (g/mol): 384.84
Molecular Formula: C12H36O4Si5
InChi Key: FBZANXDWQAVSTQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results