Learn More
DNA Ligase III Antibody, Novus Biologicals™
Product Code. 18215090 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215090 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18215090 Supplier Novus Biologicals Supplier No. NB10081661
  • ONLINE EXCLUSIVE PRICE
    583.00 EUR
    551.25 EUR / 0.1mL
    Save 31.75 EUR
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Rabbit Polyclonal Antibody
DNA Ligase III Polyclonal specifically detects DNA Ligase III in Human samples. It is validated for Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen DNA Ligase III
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P49916
Gene Alias DNA ligase 3, DNA ligase III, EC 6.5.1.1, LIG2, ligase II, DNA, ATP-dependent, ligase III, DNA, ATP-dependent, Polydeoxyribonucleotide synthase [ATP] 3
Gene Symbols LIG3
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 800 and 850 of human DNA Ligase III using the numbering given in entry CAA59230.1 (GeneID 3980).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 3980
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
For Research Use Only