missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DLL1 Monoclonal specifically detects DLL1 in Human samples. It is validated for ELISA/Western Blot.
Specifications
Specifications
| Antigen | DLL1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | D1L165-6 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Delta, delta (Drosophila)-like 1, Delta1, delta-like 1 (Drosophila), delta-like protein 1, DL1, Drosophila Delta homolog 1, H-Delta-1 |
| Host Species | Mouse |
| Immunogen | Recombinant human DLL1 |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?