missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Neutralization.
Specifications
Specifications
| Antigen | Dkk-1 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Clone | R015 |
| Conjugate | Unconjugated |
| Formulation | 0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 |
| Gene Alias | dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Dkk-1 Protein |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?