Learn More
Cytiva Disposable PD-10 Columns
Product Code. 11862200 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11862200 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11862200 Supplier Cytiva Supplier No. 17043501
In Stock
Use for gravity flow purification. Disposable PD-10 Columns are economical, plastic columns that can be packed with a wide range of bulk media.
  • Economical and versatile columns for gravity flow applications
  • Empty PD-10 columns can be packed with a wide range of bulk media
  • Easy to use
  • Use PD-10 Columns with LabMate Buffer Reservoir for convenient equilibration, sample application, and wash
  • Empty Disposable PD-10 Columns are economical, plastic columns for single-use applications using gravity flow
  • The columns hold up to approx. 13 ml of chromatography media; the frits can be used as an upper bed support that prevents the medium from running dry under gravity flow
  • Comes with caps
TRUSTED_SUSTAINABILITY
Specifications
Product Type Empty Columns, with Caps
Diameter (English) Inner Column 0.59 in.
For Use With (Application) Desalting
Diameter (Metric) Inner Column 1.5 cm
Quantity 50 Pk.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.