missing translation for 'onlineSavingsMsg'
Learn More

Diiodoacetic Acid, TRC

Product Code. 30136487
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136487 1 g 1g
17918694 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136487 Supplier Toronto Research Chemicals Supplier No. TRCD4465351G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-89-0
Molecular Formula C2 H2 I2 O2
Molecular Weight (g/mol) 311.84
Synonym 2,2-Diiodoacetic acid (ACI), Acetic acid, diiodo- (6CI, 9CI), Diiodoacetic acid
IUPAC Name 2,2-diiodoacetic acid
SMILES OC(=O)C(I)I

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H2I2O2/c3-1(4)2(5)6/h1H,(H,5,6)
Formula Weight 311.8144
Percent Purity >95%
Chemical Name or Material Diiodoacetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.