CAS RN 598-89-0
CAS RN 598-89-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diiodoacetic Acid, TRC
CAS: 598-89-0 Molecular Formula: C2 H2 I2 O2 Molecular Weight (g/mol): 311.84 Synonym: 2,2-Diiodoacetic acid (ACI),Acetic acid, diiodo- (6CI, 9CI),Diiodoacetic acid IUPAC Name: 2,2-diiodoacetic acid SMILES: OC(=O)C(I)I