missing translation for 'onlineSavingsMsg'
Learn More

Dihydrozeatin, TRC

Product Code. 30009505
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009505 5 mg 5mg
30004574 1 mg 1mg
30007617 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009505 Supplier Toronto Research Chemicals Supplier No. TRCD4548105MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14894-18-9
Molecular Formula C10H15N5O
Molecular Weight (g/mol) 221.26
Synonym Racemic Dihydrozeatin, (±)-Dihydrozeatin, 6-(4-Hydroxy-3-methylbutylamino)purine, (±)-6-(4-Hydroxy-3-methylbutylamino)purine, 2-Methyl-4-(9H-purin-6-ylamino)-1-butanol, 2-Methyl-4-(purin-6-ylamino)-1-butanol, 2-Methyl-4-(1H-purin-6-ylamino)-1-butanol;
SMILES OCC(C)CCNC1=C(NC=N2)C2=NC=N1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H15N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h5-7,16H,2-4H2,1H3,(H2,11,12,13,14,15)
Chemical Name or Material Dihydrozeatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.