CAS RN 14894-18-9
CAS RN 14894-18-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydrozeatin, TRC
CAS: 14894-18-9 Molecular Formula: C10H15N5O Molecular Weight (g/mol): 221.26 Synonym: Racemic Dihydrozeatin,(±)-Dihydrozeatin,6-(4-Hydroxy-3-methylbutylamino)purine,(±)-6-(4-Hydroxy-3-methylbutylamino)purine,2-Methyl-4-(9H-purin-6-ylamino)-1-butanol,2-Methyl-4-(purin-6-ylamino)-1-butanol,2-Methyl-4-(1H-purin-6-ylamino)-1-butanol; SMILES: OCC(C)CCNC1=C(NC=N2)C2=NC=N1