missing translation for 'onlineSavingsMsg'
Learn More

Dihydrosafrole, TRC

Product Code. 30009098
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009098 10 mg 10mg
30006838 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009098 Supplier Toronto Research Chemicals Supplier No. TRCD44992010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-58-6
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym 1,3-Benzodioxole, 5-propyl-, Benzene, 1,2-(methylenedioxy)-4-propyl- (6CI,7CI,8CI), 5-Propyl-1,3-benzodioxole, Safrole, dihydro- (6CI), 1,2-(Methylenedioxy)-4-propylbenzene, 2',3'-Dihydrosafrole, 5-Propylbenz[1,3]dioxole, Dihydrosafrol, Dihydrosafrole, NSC 27867, NSC 33707
IUPAC Name 5-propyl-1,3-benzodioxole
SMILES CCCc1ccc2OCOc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H12O2/c1-2-3-8-4-5-9-10(6-8)12-7-11-9/h4-6H,2-3,7H2,1H3
Formula Weight 164.0837
Percent Purity >95%
Chemical Name or Material Dihydrosafrole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.