CAS RN 94-58-6
CAS RN 94-58-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Dihydrosafrole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dihydrosafrole, TRC
CAS: 94-58-6 Molecular Formula: C10 H12 O2 Molecular Weight (g/mol): 164.2 Synonym: 1,3-Benzodioxole, 5-propyl-,Benzene, 1,2-(methylenedioxy)-4-propyl- (6CI,7CI,8CI),5-Propyl-1,3-benzodioxole,Safrole, dihydro- (6CI),1,2-(Methylenedioxy)-4-propylbenzene,2',3'-Dihydrosafrole,5-Propylbenz[1,3]dioxole,Dihydrosafrol,Dihydrosafrole,NSC 27867,NSC 33707 IUPAC Name: 5-propyl-1,3-benzodioxole SMILES: CCCc1ccc2OCOc2c1