Learn More
Dichlorophenylphosphine 97%, Thermo Scientific Chemicals
Product Code. 10294230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10294230 100 mL Glass bottle 100mL
10345890 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10294230 Supplier Thermo Scientific Acros Supplier No. 113661000
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 644-97-3
Molecular Formula C6H5Cl2P
Molecular Weight (g/mol) 178.98
MDL Number MFCD00000528
InChI Key IMDXZWRLUZPMDH-UHFFFAOYSA-N
Synonym dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
PubChem CID 12573
IUPAC Name dichloro(phenyl)phosphane
SMILES ClP(Cl)C1=CC=CC=C1
Specifications
Color Colorless to Yellow
Density 1.3190g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5PCl2
Refractive Index 1.5970 to 1.5990 (20°C, 589nm)
Quantity 100 mL
Beilstein 16, 763
Formula Weight 178.99
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Dichlorophenylphosphine, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorophenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H250-Catches fire spontaneously if exposed to air.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.