accessibility menu, dialog, popup

UserName

ClPCl

CAS RN 644-97-3

IUPAC Name: dichloro(phenyl)phosphane
Synonyms: dichlorophenylphosphine
Molecular Weight (g/mol): 178.98
Molecular Formula: C6H5Cl2P
InChi Key: IMDXZWRLUZPMDH-UHFFFAOYSA-N
SMILES: ClP(Cl)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)