missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetic Acid, TRC

Product Code. 17943095
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943095 250 mg 250mg
17914295 100 mg 100mg
17913745 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943095 Supplier Toronto Research Chemicals Supplier No. TRCD418730250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-64-1
Molecular Formula C2 H2 Br2 O2
Molecular Weight (g/mol) 217.84
Synonym Acetic acid, dibromo- (6CI,7CI,8CI,9CI), 2,2-Dibromoacetic acid, DBAA, Dibromoacetic acid
IUPAC Name 2,2-dibromoacetic acid
SMILES OC(=O)C(Br)Br

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2H2Br2O2/c3-1(4)2(5)6/h1H,(H,5,6)
Formula Weight 215.8422
Chemical Name or Material Dibromoacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.