CAS RN 631-64-1
CAS RN 631-64-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibromoacetic Acid, TRC
CAS: 631-64-1 Molecular Formula: C2 H2 Br2 O2 Molecular Weight (g/mol): 217.84 Synonym: Acetic acid, dibromo- (6CI,7CI,8CI,9CI),2,2-Dibromoacetic acid,DBAA,Dibromoacetic acid IUPAC Name: 2,2-dibromoacetic acid SMILES: OC(=O)C(Br)Br