Learn More
Dibenzo[a,i]pyrene, TRC
Product Code. 30007528
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007528 5 mg 5mg
30009314 100 mg 100mg
17952705 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007528 Supplier Toronto Research Chemicals Supplier No. TRCD4169855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 189-55-9
Molecular Formula C24 H14
Molecular Weight (g/mol) 302.37
Synonym Benzo[rst]pentaphene, Benzo[rst]pentacene (7CI), 1,2:7,8-Dibenzpyrene, 3,4:9,10-Dibenzopyrene, Dibenzo[a,i]pyrene, Dibenzo[b,h]pyrene, NSC 87521, Dibenz[a,i]pyrene
IUPAC Name Dibenzo[a,i]pyrene
SMILES c1ccc2c(c1)cc3ccc4cc5ccccc5c6ccc2c3c46
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C24H14/c1-3-7-19-15(5-1)13-17-9-10-18-14-16-6-2-4-8-20(16)22-12-11-21(19)23(17)24(18)22/h1-14H
Formula Weight 302.1096
Percent Purity >95%
Chemical Name or Material Dibenzo(A,I)Pyrene
Grade HPLC