CAS RN 189-55-9
CAS RN 189-55-9
IUPAC Name:
hexacyclo[10.10.2.0²,⁷.0⁹,²³.0¹⁴,¹⁹.0²⁰,²⁴]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Synonyms:
dibenzo(A,I)pyrene
Molecular Weight (g/mol):
302.38
Molecular Formula:
C24H14
InChi Key:
TUGYIJVAYAHHHM-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C=CC3=CC4=CC=CC=C4C4=CC=C2C1=C34
1
–
4
of
4
results
1
Dibenzo[a,i]pyrene, TRC
CAS: 189-55-9 Molecular Formula: C24 H14 Molecular Weight (g/mol): 302.37 Synonym: Benzo[rst]pentaphene,Benzo[rst]pentacene (7CI),1,2:7,8-Dibenzpyrene,3,4:9,10-Dibenzopyrene,Dibenzo[a,i]pyrene,Dibenzo[b,h]pyrene,NSC 87521,Dibenz[a,i]pyrene IUPAC Name: Dibenzo[a,i]pyrene SMILES: c1ccc2c(c1)cc3ccc4cc5ccccc5c6ccc2c3c46
2
Dibenzo[a,i]pyrene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality