missing translation for 'onlineSavingsMsg'
Learn More

Diazolidinyl urea, MP Biomedicals™

Product Code. 11491611 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491611 25 g 25g
11401621 100 g 100g
11411621 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491611 Supplier MP Biomedicals Supplier No. 0219019125
  • ONLINE EXCLUSIVE PRICE
    66.15 EUR
    63.00 EUR / 25g
    Save 3.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 78491-02-8
Molecular Formula C8H14N4O7
Molecular Weight (g/mol) 278.221
InChI Key SOROIESOUPGGFO-UHFFFAOYSA-N
Synonym diazolidinylurea, diazolidinyl urea, 1-1,3-bis hydroxymethyl-2,5-dioxoimidazolidin-4-yl-1,3-bis hydroxymethyl urea, germall ii, germall 11, imidazolidinyl urea 11, n,n'-bis hydroxymethyl urea, dsstox_cid_9559, dsstox_rid_78776
PubChem CID 62277
IUPAC Name 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
SMILES C(NC(=O)N(CO)C1C(=O)N(C(=O)N1CO)CO)O

Specifications

Color White
Quantity 25 g
Formula Weight 278.2
Physical Form Powder
Chemical Name or Material Diazolidinyl urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.