accessibility menu, dialog, popup

UserName

HONHONHONHOONOHO

CAS RN 78491-02-8

IUPAC Name: 1-[1,3-bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1,3-bis(hydroxymethyl)urea
Synonyms: diazolidinyl urea
Molecular Weight (g/mol): 278.22
Molecular Formula: C8H14N4O7
InChi Key: SOROIESOUPGGFO-UHFFFAOYNA-N
SMILES: OCNC(=O)N(CO)C1N(CO)C(=O)N(CO)C1=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results