missing translation for 'onlineSavingsMsg'
Learn More

Diamino-5-phenylthiazole Hydrobromide, TRC

Product Code. 30138516
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138516 5 g 5g
30009960 250 mg 250mg
30005514 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138516 Supplier Toronto Research Chemicals Supplier No. TRCD4165905G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6020-54-8
Molecular Formula C9 H9 N3 S . Br H
Molecular Weight (g/mol) 272.16
IUPAC Name 5-phenyl-1,3-thiazole-2,4-diamine;hydrobromide
SMILES Br.Nc1nc(N)c(s1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H9N3S.BrH/c10-8-7(13-9(11)12-8)6-4-2-1-3-5-6;/h1-5H,10H2,(H2,11,12);1H
Formula Weight 270.9779
Chemical Name or Material Diamino-5-phenylthiazole Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.