CAS RN 6020-54-8
CAS RN 6020-54-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diamino-5-phenylthiazole Hydrobromide, TRC
CAS: 6020-54-8 Molecular Formula: C9 H9 N3 S . Br H Molecular Weight (g/mol): 272.16 IUPAC Name: 5-phenyl-1,3-thiazole-2,4-diamine;hydrobromide SMILES: Br.Nc1nc(N)c(s1)c2ccccc2