missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Sodium Tartrate Dihydrate, Supelco™
Specifications
Specifications
| Quantity | 1 kg |
| MDL Number | MFCD00150035 |
| Synonym | di-Sodium tartrate dihydrate, di-Sodium tartrate dihydrate, Tartaric acid sodium salt dihydrate, di-Sodium tartrate dihydrate |
| Molecular Weight (g/mol) | 230.08 |
| Grade | Research |
| Density | 1.82 g/cm3 |
| Chemical Name or Material | Di-Sodium Tartrate Dihydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?