accessibility menu, dialog, popup

UserName

CAS RN 6106-24-7

IUPAC Name: 2,3-dihydroxybutanedioic acid dihydrate disodium
Synonyms: (.+-.)-tartaric acid dihydrate disodium
Molecular Weight (g/mol): 232.1
Molecular Formula: C4H10Na2O8
InChi Key: YAPIZISBZUXUIH-UHFFFAOYNA-N
SMILES: O.O.[Na].[Na].OC(C(O)C(O)=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (20)
  • (1)
Grade 
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (20)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)