missing translation for 'onlineSavingsMsg'
Learn More

di-Phosphorus Pentoxide, Supelco™

Product Code. 15823920
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15823920 500 g 500g
15813920 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15823920 Supplier Supelco Supplier No. 1005700500
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis ACS,ISO,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3

Specifications

Melting Point 420°C (closed capillary tube)
Quantity 500 g
MDL Number MFCD00148880
Synonym di-Phosphorus pentoxide, Phosphoric anhydride, di-Phosphorus pentoxide
Molecular Weight (g/mol) 141.94
Grade Research
Density 2.28 g/cm3 at 20 to 25°C
Chemical Name or Material Di-Phosphorus Pentoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.