accessibility menu, dialog, popup

UserName

OPOPOOPOOOPOOO

CAS RN 1314-56-3

IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Synonyms: phosphorus pentoxide diphosphorus pentoxide tetraphosphorus decaoxide phosphor(V)-oxid phosphoric anhydride phosphorus(V) oxide phosphorpentoxid
Molecular Weight (g/mol): 283.89
Molecular Formula: O10P4
InChi Key: DLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILES: O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Quantity 
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
Physical Form 
  • (4)
  • (12)
  • (5)
  • (3)
Grade 
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (3)
  • (9)
  • (11)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (4)
  • (12)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)