missing translation for 'onlineSavingsMsg'
Learn More
Learn More
di-Phosphorus Pentasulfide, Sigma-Aldrich™
Specifications
Specifications
| Boiling Point | 515°C |
| Melting Point | 280°C to 284°C (lit.) |
| Quantity | 1 kg |
| MDL Number | MFCD00011441 |
| Synonym | Phosphorus pentasulfide, Diphosphorus pentasulfide, Phosphorus(V) sulfide, Phosphorus(V) sulfide, Phosphorus pentasulfide, Phosphorus(V) sulfide |
| Molecular Weight (g/mol) | 222.27 |
| Grade | Research |
| Density | 2.09 g/mL at 25°C |
| Chemical Name or Material | Di-Phosphorus Pentasulfide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?