accessibility menu, dialog, popup

UserName

SPSPSSPSSSPSSS

CAS RN 1314-80-3

IUPAC Name: tricyclo[3.3.1.1³,⁷]tetraphosphathiane-1,3,5,7-tetrathione
Synonyms: phosphorus pentasulfide
Molecular Weight (g/mol): 444.5
Molecular Formula: P4S10
InChi Key: CYQAYERJWZKYML-UHFFFAOYSA-N
SMILES: S=P12SP3(=S)SP(=S)(S1)SP(=S)(S2)S3
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)