missing translation for 'onlineSavingsMsg'
Learn More

Desethyl Terbuthylazine, TRC

Product Code. 30132661
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132661 500 mg 500mg
30136739 1 g 1g
18000060 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132661 Supplier Toronto Research Chemicals Supplier No. TRCD289700500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30125-63-4
Molecular Formula C7 H12 Cl N5
Molecular Weight (g/mol) 201.66
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-(1,1-dimethylethyl)-, 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(1,1-dimethylethyl)- (9CI), s-Triazine, 2-amino-4-(tert-butylamino)-6-chloro- (8CI), 6-Chloro-N2-(1,1-dimethylethyl)-1,3,5-triazine-2,4-diamine, 2-Chloro-4-amino-6-tert-butylamino-1,3,5-triazine, 2-Chloro-4-amino-6-tert-butylamino-s-triazine, Deethylterbuthylazine, Deethylterbutylazine, Desethylterbuthylazine, Desethylterbutylaszine, Desethylterbutylazine, GS 26379, Terbuthylazine-desethyl, Terbutylazine desethyl
IUPAC Name 4-N-tert-butyl-6-chloro-1,3,5-triazine-2,4-diamine
SMILES CC(C)(C)Nc1nc(N)nc(Cl)n1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H12ClN5/c1-7(2,3)13-6-11-4(8)10-5(9)12-6/h1-3H3,(H3,9,10,11,12,13)
Formula Weight 201.0781
Percent Purity >95%
Chemical Name or Material Terbuthylazine-desethyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.