accessibility menu, dialog, popup

UserName

CAS RN 30125-63-4

H3CH3CCH3NHNNH2NClN

CAS RN 30125-63-4

IUPAC Name: N2-tert-butyl-6-chloro-1,3,5-triazine-2,4-diamine
Synonyms: terbuthylazine-desethyl
Molecular Weight (g/mol): 201.66
Molecular Formula: C7H12ClN5
InChi Key: LMKQNTMFZLAJDV-UHFFFAOYSA-N
SMILES: CC(C)(C)NC1=NC(N)=NC(Cl)=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)
  • (1)
Solvent or Matrix 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results