missing translation for 'onlineSavingsMsg'
Learn More

Deschloro-4,4’-dichloro Clomiphene Citrate (E/Z Mixture), TRC

Product Code. 30136974
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136974 500 mg 500mg
30008678 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136974 Supplier Toronto Research Chemicals Supplier No. TRCD289020500MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117884-83-0
Molecular Formula C26 H27 Cl2 N O . C6 H8 O7
Molecular Weight (g/mol) 632.53
Synonym 2-[4-[1,2-bis(4-chlorophenyl)ethenyl]phenoxy]-N,N-diethylethanamine 2-Hydroxy-1,2,3-propanetricarboxylate (1:1), 2-[p-[1,2-bis(p-chlorophenyl)vinyl]phenoxy]triethylamine citrate (6CI)
IUPAC Name 2-[4-[1,2-bis(4-chlorophenyl)ethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOc1ccc(cc1)C(=Cc2ccc(Cl)cc2)c3ccc(Cl)cc3.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C26H27Cl2NO.C6H8O7/c1-3-29(4-2)17-18-30-25-15-9-22(10-16-25)26(21-7-13-24(28)14-8-21)19-20-5-11-23(27)12-6-20;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-16,19H,3-4,17-18H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 631.174
Chemical Name or Material Deschloro-4,4'-dichloro Clomiphene Citrate (EZ mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.