CAS RN 117884-83-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deschloro-4,4’-dichloro Clomiphene Citrate (E/Z Mixture), TRC
CAS: 117884-83-0 Molecular Formula: C26 H27 Cl2 N O . C6 H8 O7 Molecular Weight (g/mol): 632.53 Synonym: 2-[4-[1,2-bis(4-chlorophenyl)ethenyl]phenoxy]-N,N-diethylethanamine 2-Hydroxy-1,2,3-propanetricarboxylate (1:1),2-[p-[1,2-bis(p-chlorophenyl)vinyl]phenoxy]triethylamine citrate (6CI) IUPAC Name: 2-[4-[1,2-bis(4-chlorophenyl)ethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: CCN(CC)CCOc1ccc(cc1)C(=Cc2ccc(Cl)cc2)c3ccc(Cl)cc3.OC(=O)CC(O)(CC(=O)O)C(=O)O