missing translation for 'onlineSavingsMsg'
Learn More

Deethylphosphamidon, TRC

Product Code. 30134937
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134937 100 mg 100mg
30003857 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134937 Supplier Toronto Research Chemicals Supplier No. TRCD228580100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13171-22-7
Molecular Formula C8H15ClNO5P
Molecular Weight (g/mol) 271.64
Synonym 2-Chloro-3-(ethylamino)-1-methyl-3-oxo-1-propenyl Dimethyl Ester Phosphoric Acid, Phosphoric Acid Dimethyl Ester, Ester with 2-Chloro-N-ethyl-3-hydroxycrotonamide, 2-Chloro-N-ethyl-3-hydroxycrotonamide Dimethyl Phosphate (Ester), Desethylphosphamidon, Dimethyl Phosphate, Ester with 2-Chloro-N-ethyl-3-hydroxycrotonamide, N-Deethylphosphamidon
IUPAC Name (Z)-3-chloro-4-(ethylamino)-4-oxobut-2-en-2-yl dimethyl phosphate
SMILES O=C(/C(Cl)=C(C)/OP(OC)(OC)=O)NCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H15ClNO5P/c1-5-10-8(11)7(9)6(2)15-16(12,13-3)14-4/h5H2,1-4H3,(H,10,11)/b7-6-
Formula Weight 271.04
Chemical Name or Material Deethylphosphamidon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.