CAS RN 13171-22-7
CAS RN 13171-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deethylphosphamidon, TRC
CAS: 13171-22-7 Molecular Formula: C8H15ClNO5P Molecular Weight (g/mol): 271.64 Synonym: 2-Chloro-3-(ethylamino)-1-methyl-3-oxo-1-propenyl Dimethyl Ester Phosphoric Acid,Phosphoric Acid Dimethyl Ester, Ester with 2-Chloro-N-ethyl-3-hydroxycrotonamide,2-Chloro-N-ethyl-3-hydroxycrotonamide Dimethyl Phosphate (Ester),Desethylphosphamidon,Dimethyl Phosphate, Ester with 2-Chloro-N-ethyl-3-hydroxycrotonamide,N-Deethylphosphamidon IUPAC Name: (Z)-3-chloro-4-(ethylamino)-4-oxobut-2-en-2-yl dimethyl phosphate SMILES: O=C(/C(Cl)=C(C)/OP(OC)(OC)=O)NCC