missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, Sigma-Aldrich™

Product Code. 15898920
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 kg
Unit Size:
1 L
100mL
2.5 L
25kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898920 100 mL 100mL
15846340 1 L 1 L
15808930 2.5 L 2.5 L
15856340 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15898920 Supplier Sigma Aldrich Supplier No. 8031010100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mixture of cis-and trans isomers for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8

Specifications

Melting Point -40°C
Density 0.88 g/cm3 at 20°C
Boiling Point 189°C to 191°C/1013 hPa
MDL Number MFCD00004130
Quantity 100 mL
Synonym Decahydronaphthalene, Decalin, Decahydronaphthalene
Molecular Weight (g/mol) 138.25
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.