accessibility menu, dialog, popup

UserName

CAS RN 91-17-8

IUPAC Name: decahydronaphthalene
Synonyms: decalin (cis)-isomer decalin DEC decahydronaphthalene (substance) (trans)-isomer decalin decalin, (trans)- decalin, (cis)- de-kalin dekahydronaphthalin dekalin
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: NNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILES: C1CCC2CCCCC2C1
Quantity 
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (14)
  • (8)
Boiling Point 
  • (1)
  • (7)
  • (4)
  • (4)
Percent Purity 
  • (4)
  • (9)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (6)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (14)
  • (8)
  • (1)
  • (7)
  • (4)
  • (4)
  • (4)
  • (9)
  • (2)
  • (8)
  • (4)